The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289654.1 | NP_001276583.1 | 23 - 24 | 5027 | 79 |
NM_001289655.1 | NP_001276584.1 | 24 - 25 | 5384 | 79 | |
NM_175276.4 | NP_780485.2 | 25 - 26 | 5480 | 79 | |
GenBank mRNA | AB078608.1 | - | 25 - 26 | 4658 | 79 |
AB078609.1 | - | 23 - 24 | 4205 | 79 | |
AB100088.1 | - | 24 - 25 | 4562 | 79 | |
AK220413.1 | - | 21 - 22 | 3644 | 79 | |
BC056371.1 | - | 15 - 16 | 3110 | 79 | |
BC060607.1 | - | 10 - 11 | 2436 | 79 | |
BC138046.1 | - | 23 - 24 | 4127 | 79 |