The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009486.3 | NP_033512.1 | 3 - 4 | 1375 | 163 |
NR_104384.1 | 2 - 3 | 166 | |||
GenBank mRNA | AF011420.1 | - | 3 - 4 | 1374 | 163 |
BC114970.1 | - | 2 - 3 | 166 |