The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008936.1 | NP_032962.1 | 1 - 2 | 339 | 67 |
GenBank mRNA | U77946.1 | - | 1 - 2 | 339 | 67 |