The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145827.3 | NP_665826.1 | 2 - 3 | 499 | 84 |
GenBank mRNA | AF486632.1 | - | 3 - 4 | 395 | 84 |
AY337285.1 | - | 1 - 2 | 285 | 84 | |
AY355340.1 | - | 1 - 2 | 273 | 84 | |
AY495376.1 | - | 2 - 3 | 499 | 84 | |
AY495377.1 | - | 3 - 4 | 395 | 84 | |
BC116174.1 | - | 2 - 3 | 459 | 84 | |
BC116175.1 | - | 2 - 3 | 459 | 84 | |
KF032621.1 | - | 1 - 2 | 275 | 84 |