The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020277.2 | NP_064673.2 | 16 - 17 | 2440 | 56 |
GenBank mRNA | AB039952.1 | - | 15 - 16 | 2357 | 56 |
AF228681.1 | - | 16 - 17 | 2440 | 56 | |
AJ271092.2 | - | 15 - 16 | 2353 | 56 | |
AK035197.1 | - | 12 - 13 | 1970 | 56 | |
AK044908.1 | - | 12 - 13 | 2055 | 56 | |
AK161030.1 | - | 7 - 8 | 1033 | 56 | |
AY280364.1 | - | 16 - 17 | 2404 | 56 | |
AY280365.1 | - | 16 - 17 | 2404 | 56 | |
BC133712.1 | - | 15 - 16 | 2357 | 56 |