Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hgf
Entrez Gene ID
15234
Gene Name
hepatocyte growth factor
Gene Aliases
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF
Chromosome Location
Chr.5: 16553335 - 16622408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289458.1 NP_001276387.1 4 - 5 564 128
NM_001289459.1 NP_001276388.1 4 - 5 553 128
NM_001289460.1 NP_001276389.1 4 - 5 564 128
NM_001289461.1 NP_001276390.1 3 - 4 386 128
NM_010427.5 NP_034557.3 4 - 5 536 128
GenBank mRNA AF042856.1 - 3 - 4 365 128
AK042121.1 - 4 - 5 482 128
AK052289.1 - 4 - 5 565 128
AK082461.1 - 3 - 4 536 128
AK142159.1 - 4 - 5 482 128
BC119228.1 - 4 - 5 423 128
BC144764.1 - 4 - 5 435 129
D10212.1 - 3 - 4 359 128
D10213.1 - 3 - 4 359 128
S71816.1 - 3 - 4 359 128
X72307.1 - 3 - 4 420 128
X84046.1 - 3 - 4 399 128

View Full Product Details