Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bsg
Entrez Gene ID
12215
Gene Name
basigin
Gene Aliases
AI115436, AI325119, CD147, EMMPRIN, HT-7
Chromosome Location
Chr.10: 79704358 - 79711979 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077184.1 NP_001070652.1 6 - 7 917 70
NM_009768.2 NP_033898.1 7 - 8 1265 70
GenBank mRNA AK002332.1 - 6 - 7 769 70
AK011364.1 - 6 - 7 768 70
AK147225.1 - 6 - 7 5591 70
AY089967.1 - 7 - 8 1123 70
BC010270.1 - 6 - 7 760 70
D00611.1 - 6 - 7 767 70
S63813.1 - 6 - 7 767 70
Y16256.1 - 6 - 7 767 70
Y16257.1 - 5 - 6 763 70
Y16258.1 - 5 - 6 709 70

View Full Product Details