The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039185.1 | NP_001034274.1 | 2 - 3 | 512 | 61 |
NM_001039186.1 | NP_001034275.1 | 2 - 3 | 512 | 61 | |
GenBank mRNA | AB236330.2 | - | 2 - 3 | 421 | 61 |
AB236332.2 | - | 2 - 3 | 421 | 61 | |
AK085749.1 | - | 2 - 3 | 472 | 61 | |
BC016891.1 | - | 2 - 3 | 464 | 61 | |
BC121189.1 | - | 1 - 2 | 69 | 61 | |
M77196.1 | - | 2 - 3 | 490 | 61 | |
X15351.1 | - | 2 - 3 | 524 | 61 | |
X67279.1 | - | 2 - 3 | 524 | 61 |