Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hk1
Entrez Gene ID
15275
Gene Name
hexokinase 1
Gene Aliases
BB404130, Hk-1, Hk1-s, dea, mHk1-s
Chromosome Location
Chr.10: 62268855 - 62379920 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146100.1 NP_001139572.1 15 - 16 2565 76
NM_010438.3 NP_034568.2 19 - 20 2508 76
GenBank mRNA AK143630.1 - 15 - 16 2503 76
AK161466.1 - 9 - 10 1477 76
AK167468.1 - 15 - 16 2321 76
AK197700.1 - 1 - 2 145 76
AK212693.1 - 1 - 2 144 76
AK218898.1 - 1 - 2 144 76
BC027316.1 - 13 - 14 1660 76
BC072628.1 - 15 - 16 2686 76
J05277.1 - 15 - 16 2223 76
L16949.1 - 19 - 20 2487 76

View Full Product Details