The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024837.2 | NP_001020008.1 | 1 - 2 | 177 | 86 |
NM_130895.3 | NP_570965.2 | 1 - 2 | 177 | 86 | |
NR_004429.1 | 1 - 2 | 177 | 86 | ||
NR_021486.1 | 1 - 2 | 177 | 86 | ||
GenBank mRNA | AF403106.2 | - | 1 - 2 | 129 | 86 |
AF403107.2 | - | 1 - 2 | 129 | 86 | |
AF403108.2 | - | 1 - 2 | 129 | 86 | |
AF403109.2 | - | 1 - 2 | 129 | 86 | |
AF525421.1 | - | 1 - 2 | 129 | 86 | |
AK138318.1 | - | 1 - 2 | 170 | 86 | |
AK141777.1 | - | 1 - 2 | 149 | 86 | |
AK164931.1 | - | 1 - 2 | 145 | 86 | |
AY162454.1 | - | 1 - 2 | 129 | 86 | |
BB622419.1 | - | 1 - 2 | 177 | 86 | |
BC051107.1 | - | 1 - 2 | 115 | 86 | |
BC062806.1 | - | 1 - 2 | 132 | 86 | |
BF017513.1 | - | 2 - 3 | 86 | ||
CJ115965.1 | - | 1 - 2 | 177 | 86 |