Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan15
Entrez Gene ID
70423
Gene Name
tetraspanin 15
Gene Aliases
1110036D12Rik, 2700063A19Rik, AW048364, Tm4sf15
Chromosome Location
Chr.10: 62185396 - 62231277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197996.2 NP_932113.2 2 - 3 381 107
GenBank mRNA AK012471.1 - 2 - 3 380 107
AK172169.1 - 2 - 3 391 107
BC003872.1 - 2 - 3 330 107
CN455615.1 - 2 - 3 270 107
CX206586.1 - 2 - 3 381 107

View Full Product Details