Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf2
Entrez Gene ID
74766
Gene Name
Yip1 domain family, member 2
Gene Aliases
1300010K09Rik
Chromosome Location
Chr.9: 21588682 - 21592831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205157.1 NP_001192086.1 8 - 9 961 61
NM_001205158.1 NP_001192087.1 8 - 9 951 61
NM_138303.2 NP_612176.1 7 - 8 1099 61
NR_038062.1 7 - 8 900 61
GenBank mRNA AK004959.1 - 7 - 8 885 61
AK033705.1 - 8 - 9 910 61
AK133631.1 - 8 - 9 921 61
BC002282.1 - 7 - 8 1014 61
BC003289.1 - 8 - 9 892 61

View Full Product Details