Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf2
Entrez Gene ID
74766
Gene Name
Yip1 domain family, member 2
Gene Aliases
1300010K09Rik
Chromosome Location
Chr.9: 21588682 - 21592831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205158.1 NP_001192087.1 9 - 10 1075 68
NR_038062.1 8 - 9 1024 68
GenBank mRNA AK004959.1 - 8 - 9 1009 68
BC003289.1 - 9 - 10 1016 68

View Full Product Details