Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkcsh
Entrez Gene ID
19089
Gene Name
protein kinase C substrate 80K-H
Gene Aliases
80K-H, PKCSH
Chromosome Location
Chr.9: 22002988 - 22014245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293650.1 NP_001280579.1 4 - 5 455 57
NM_001293651.1 NP_001280580.1 4 - 5 459 57
NM_008925.2 NP_032951.1 4 - 5 455 57
GenBank mRNA AK137661.1 - 4 - 5 422 57
AK153931.1 - 4 - 5 430 57
AK159705.1 - 4 - 5 426 57
BC009816.1 - 4 - 5 388 57
CD807078.1 - 4 - 5 424 57
U92794.1 - 4 - 5 408 57

View Full Product Details