Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd1a
Entrez Gene ID
52857
Gene Name
GRAM domain containing 1A
Gene Aliases
1300003M23Rik, AI415002, AW547240, D7Bwg0611e
Chromosome Location
Chr.7: 31130127 - 31155935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027898.3 NP_082174.3 7 - 8 765 70
GenBank mRNA AK004885.1 - 7 - 8 751 70
AK083837.1 - 7 - 8 728 70
AK150032.1 - 7 - 8 704 70
AK150196.1 - 7 - 8 717 70
AK150837.1 - 7 - 8 731 70
AK151987.1 - 7 - 8 731 70
AK152294.1 - 7 - 8 731 70
AK154821.1 - 7 - 8 766 70
AK205495.1 - 3 - 4 252 70

View Full Product Details