The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252450.1 | NP_001239379.1 | 5 - 6 | 777 | 54 |
NM_171826.3 | NP_741968.1 | 5 - 6 | 784 | 54 | |
NR_045517.1 | 4 - 5 | 733 | 54 | ||
GenBank mRNA | AK003808.1 | - | 4 - 5 | 625 | 54 |
AK009432.1 | - | 4 - 5 | 597 | 54 | |
AK009743.1 | - | 4 - 5 | 617 | 54 | |
AK012260.1 | - | 4 - 5 | 646 | 54 | |
AK152121.1 | - | 4 - 5 | 628 | 54 | |
AK152722.1 | - | 4 - 5 | 623 | 54 | |
BC055695.1 | - | 3 - 4 | 541 | 54 | |
CA750661.1 | - | 5 - 6 | 627 | 54 |