Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocos
Entrez Gene ID
68591
Gene Name
molybdenum cofactor sulfurase
Gene Aliases
1110018O12Rik, HMCS, MCS, MOS
Chromosome Location
Chr.18: 24653176 - 24701577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026779.1 NP_081055.1 7 - 8 1571 63
GenBank mRNA AK076385.1 - 7 - 8 1385 63
BC016223.1 - 7 - 8 1386 63
BC113180.1 - 7 - 8 1571 63
BC113786.1 - 7 - 8 1571 63

View Full Product Details