The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025364.2 | NP_079640.1 | 9 - 10 | 668 | 85 |
GenBank mRNA | AK003453.1 | - | 9 - 10 | 520 | 85 |
AK018773.1 | - | 9 - 10 | 531 | 85 | |
AK088266.1 | - | 9 - 10 | 517 | 85 | |
AK146248.1 | - | 9 - 10 | 546 | 85 | |
BC027510.1 | - | 9 - 10 | 507 | 85 | |
BC094393.1 | - | 10 - 11 | 699 | 85 | |
BC157909.1 | - | 9 - 10 | 532 | 85 | |
BC158023.1 | - | 9 - 10 | 532 | 85 | |
BC172063.1 | - | 9 - 10 | 529 | 85 | |
BF148013.1 | - | 2 - 3 | 85 |