The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028258.1 | 1 - 2 | 220 | 56 | |
GenBank mRNA | BC172157.1 | - | 1 - 2 | 220 | 56 |
CF739520.1 | - | 1 - 2 | 114 | 56 |