Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Orc5
Entrez Gene ID
26429
Gene Name
origin recognition complex, subunit 5
Gene Aliases
AL033327, MmORC5, Orc5l
Chromosome Location
Chr.5: 22486484 - 22550367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011959.2 NP_036089.1 12 - 13 1218 112
GenBank mRNA AJ007360.1 - 12 - 13 1215 112
AK084683.1 - 12 - 13 1246 112
AK144983.1 - 12 - 13 1234 112
AK165912.1 - 12 - 13 1224 112
AK166537.1 - 12 - 13 1157 112
AK169683.1 - 11 - 12 1273 112
BC006927.1 - 12 - 13 1234 112

View Full Product Details