Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnt16
Entrez Gene ID
93735
Gene Name
wingless-type MMTV integration site family, member 16
Gene Aliases
E130309I19Rik
Chromosome Location
Chr.6: 22288165 - 22298522 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053116.4 NP_444346.3 1 - 2 379 107
GenBank mRNA AK043534.1 - 1 - 2 319 107
AK053805.1 - 1 - 2 380 107
AK054110.1 - 1 - 2 377 107
BC115811.1 - 1 - 2 339 107
BC115925.1 - 1 - 2 339 107

View Full Product Details