Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec2l
Entrez Gene ID
665180
Gene Name
C-type lectin domain family 2, member L
Gene Aliases
AI414054, AW122724, Gm1064
Chromosome Location
Chr.6: 38663069 - 38680865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101507.1 NP_001094977.1 1 - 2 351 83
GenBank mRNA CJ059176.1 - 1 - 2 142 83
CJ060465.1 - 1 - 2 353 83
CJ060631.1 - 1 - 2 353 83

View Full Product Details