Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc39a3
- Entrez Gene ID
-
106947
- Gene Name
- solute carrier family 39 (zinc transporter), member 3
- Gene Aliases
-
AI845814, Zip3
- Chromosome Location
-
Chr.10: 81028540 - 81037421 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC085175.1 |
- |
1 - 2 |
28 |
60 |
View Full Product Details