Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a3
Entrez Gene ID
106947
Gene Name
solute carrier family 39 (zinc transporter), member 3
Gene Aliases
AI845814, Zip3
Chromosome Location
Chr.10: 81028540 - 81037421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK087519.1 - 2 - 3 132 76
BC085175.1 - 2 - 3 115 76

View Full Product Details