Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Adarb1
- Entrez Gene ID
-
110532
- Gene Name
- adenosine deaminase, RNA-specific, B1
- Gene Aliases
-
1700057H01Rik, AW124433, AW558573, Adar2, BB220382, D10Bwg0447e, Red1
- Chromosome Location
-
Chr.10: 77290727 - 77418273 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF525421.1 |
- |
4 - 5 |
443 |
95 |
AY162454.1 |
- |
4 - 5 |
443 |
95 |
View Full Product Details