Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm10
Entrez Gene ID
382066
Gene Name
PR domain containing 10
Gene Aliases
Gm1112, tristanin
Chromosome Location
Chr.9: 31278780 - 31381731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080817.1 NP_001074286.1 10 - 11 1710 66

View Full Product Details