Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc33
Entrez Gene ID
382077
Gene Name
coiled-coil domain containing 33
Gene Aliases
4930535E21Rik, Gm1117, Tzip1
Chromosome Location
Chr.9: 58028673 - 58118823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166282.1 NP_001159754.1 5 - 6 1133 65
NM_029212.3 NP_083488.1 3 - 4 602 65
GenBank mRNA AB048860.1 - 3 - 4 676 65
AK145264.1 - 5 - 6 1135 65
BC116291.1 - 3 - 4 602 65
BC116292.1 - 3 - 4 602 65

View Full Product Details