Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aebp1
Entrez Gene ID
11568
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr.11: 5861853 - 5872225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF053943.1 - 19 - 20 2883 57
AK004712.1 - 15 - 16 1892 57
AK128980.1 - 19 - 20 2959 57
AK158276.1 - 19 - 20 2909 57
AK159330.1 - 19 - 20 2890 57
AK159342.1 - 19 - 20 2909 57
AK159377.1 - 19 - 20 2880 57
AK159409.1 - 19 - 20 2910 57
AK159774.1 - 19 - 20 2881 57
AK159957.1 - 19 - 20 2881 57
AK216593.1 - 2 - 3 186 57
BC082577.1 - 19 - 20 2895 57
X80478.1 - 10 - 11 1613 57

View Full Product Details