The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011507.3 | NP_035637.2 | 9 - 10 | 1140 | 88 |
GenBank mRNA | AF058956.1 | - | 8 - 9 | 982 | 88 |
AK045176.1 | - | 9 - 10 | 1093 | 88 | |
AK156263.1 | - | 9 - 10 | 1096 | 88 | |
AK167136.1 | - | 9 - 10 | 1095 | 88 | |
AK167339.1 | - | 9 - 10 | 1379 | 88 | |
AK172561.1 | - | 9 - 10 | 1095 | 88 | |
AK172633.1 | - | 9 - 10 | 1095 | 88 | |
BC030947.1 | - | 1 - 2 | 117 | 88 | |
BC043312.1 | - | 7 - 8 | 835 | 88 | |
BC054425.1 | - | 8 - 9 | 893 | 88 | |
BC080781.1 | - | 9 - 10 | 1095 | 88 | |
EU234009.1 | - | 9 - 10 | 1047 | 88 |