The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081146.2 | NP_001074615.1 | 4 - 5 | 462 | 75 |
NM_001134459.1 | NP_001127931.1 | 3 - 4 | 541 | 75 | |
NM_001134460.2 | NP_001127932.1 | 3 - 4 | 863 | 75 | |
NM_001134461.1 | NP_001127933.1 | 3 - 4 | 358 | 75 | |
GenBank mRNA | AK032631.1 | - | 3 - 4 | 866 | 75 |
AK147576.1 | - | 3 - 4 | 542 | 75 | |
AK147628.1 | - | 3 - 4 | 542 | 75 | |
BC050793.1 | - | 3 - 4 | 358 | 75 | |
BC099897.1 | - | 4 - 5 | 433 | 75 | |
BC145754.1 | - | 3 - 4 | 380 | 75 |