Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmf1
Entrez Gene ID
232286
Gene Name
TATA element modulatory factor 1
Gene Aliases
7030402D04Rik, Gm153, Tmf1`
Chromosome Location
Chr.6: 97151950 - 97179124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081111.2 NP_001074580.1 16 - 17 3233 102
GenBank mRNA BC150931.1 - 16 - 17 3244 102
BQ561256.1 - 1 - 2 102

View Full Product Details