Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abtb1
Entrez Gene ID
80283
Gene Name
ankyrin repeat and BTB (POZ) domain containing 1
Gene Aliases
AI847549, BC003234, BPOZ, EF1ABP
Chromosome Location
Chr.6: 88835914 - 88842386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030251.3 NP_084527.2 8 - 9 868 64
GenBank mRNA AB053477.1 - 8 - 9 799 64
AI117801.1 - 1 - 2 39 64
AK049100.1 - 8 - 9 784 64
AK088437.1 - 8 - 9 833 64
AK155432.1 - 3 - 4 2290 64
AK155505.1 - 3 - 4 2290 64
AK155644.1 - 3 - 4 2290 64
AK220208.1 - 8 - 9 712 64
BC003234.1 - 8 - 9 812 64
CO804154.1 - 1 - 2 64

View Full Product Details