Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfpt1
Entrez Gene ID
14583
Gene Name
glutamine fructose-6-phosphate transaminase 1
Gene Aliases
2810423A18Rik, AI324119, AI449986, GFA, GFAT, GFAT1, GFAT1m, Gfpt
Chromosome Location
Chr.6: 87042846 - 87092207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013528.3 NP_038556.1 18 - 19 2157 114
GenBank mRNA AF334736.1 - 19 - 20 2162 114
AK019852.1 - 18 - 19 2153 114
BC002283.1 - 15 - 16 1756 114
BC010516.1 - 18 - 19 2105 114
BC050762.1 - 18 - 19 2153 114

View Full Product Details