Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snrnp40
Entrez Gene ID
66585
Gene Name
small nuclear ribonucleoprotein 40 (U5)
Gene Aliases
0610009C03Rik, Prp8bp, Wdr57
Chromosome Location
Chr.4: 130360132 - 130390033 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025645.2 NP_079921.2 7 - 8 936 104
GenBank mRNA AK002371.1 - 7 - 8 900 104
AK136135.1 - 7 - 8 866 104
AK152287.1 - 7 - 8 900 104
AK152352.1 - 7 - 8 894 104
AK169605.1 - 7 - 8 882 104
BC058365.1 - 7 - 8 868 104

View Full Product Details