Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cnrip1
- Entrez Gene ID
-
380686
- Gene Name
- cannabinoid receptor interacting protein 1
- Gene Aliases
-
1500041B16Rik, 3110054C06Rik, 5330437A18Rik, AI854501, C2orf32
- Chromosome Location
-
Chr.11: 17051934 - 17079372 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005381.1 |
- |
1 - 2 |
245 |
83 |
View Full Product Details