Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hbs1l
Entrez Gene ID
56422
Gene Name
Hbs1-like (S. cerevisiae)
Gene Aliases
2810035F15Rik, AI326327, eRFS
Chromosome Location
Chr.10: 21295979 - 21368889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042593.1 NP_001036058.1 16 - 17 2040 96
NM_019702.2 NP_062676.2 16 - 17 2049 96
GenBank mRNA AF087672.1 - 13 - 14 1910 96
AK146678.1 - 16 - 17 2006 96
AK166140.1 - 16 - 17 2003 96
AK168671.1 - 16 - 17 2003 96
AK169281.1 - 16 - 17 2003 96
AK173091.1 - 16 - 17 1995 96
BC010251.1 - 16 - 17 1974 96

View Full Product Details