Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsph4a
Entrez Gene ID
212892
Gene Name
radial spoke head 4 homolog A (Chlamydomonas)
Gene Aliases
A230081C05, Rshl3
Chromosome Location
Chr.10: 33905111 - 33916021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162957.1 NP_001156429.1 1 - 2 725 79
GenBank mRNA AK038980.1 - 1 - 2 352 79
BC138000.1 - 1 - 2 725 79
BC138001.1 - 1 - 2 725 79

View Full Product Details