The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178713.4 | NP_848828.1 | 1 - 2 | 180 | 154 |
GenBank mRNA | AF510322.1 | - | 1 - 2 | 189 | 154 |
AK050298.1 | - | 1 - 2 | 181 | 154 | |
AK143752.1 | - | 1 - 2 | 181 | 154 | |
AK165447.1 | - | 1 - 2 | 181 | 154 | |
BC038493.1 | - | 1 - 2 | 192 | 154 |