Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid1a
Entrez Gene ID
93760
Gene Name
AT rich interactive domain 1A (SWI-like)
Gene Aliases
1110030E03Rik, BAF250a, Osa1, Smarcf1
Chromosome Location
Chr.4: 133679008 - 133753981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080819.1 NP_001074288.1 1 - 2 1141 102

View Full Product Details