The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080819.1 | NP_001074288.1 | 6 - 7 | 2249 | 139 |
GenBank mRNA | AK142143.1 | - | 5 - 6 | 1431 | 139 |
BC082554.1 | - | - | 1481 | 139 |