The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172938.3 | NP_766526.3 | 6 - 7 | 1163 | 74 |
GenBank mRNA | AK034171.1 | - | 6 - 7 | 1002 | 74 |
AK037907.1 | - | 1 - 2 | 233 | 74 | |
AK040597.1 | - | 1 - 2 | 811 | 74 | |
AK047972.1 | - | 4 - 5 | 589 | 74 | |
AK172160.1 | - | 2 - 3 | 226 | 74 | |
AK220309.1 | - | 4 - 5 | 547 | 74 | |
BC043310.1 | - | 6 - 7 | 958 | 74 |