The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172938.3 | NP_766526.3 | 8 - 9 | 1578 | 63 |
GenBank mRNA | AF397912.1 | - | 1 - 2 | 115 | 63 |
AK034171.1 | - | 8 - 9 | 1417 | 63 | |
AK040597.1 | - | 3 - 4 | 1226 | 63 | |
AK043500.1 | - | 3 - 4 | 1109 | 63 | |
AK047972.1 | - | 6 - 7 | 1004 | 63 | |
AK141826.1 | - | 3 - 4 | 602 | 63 | |
AK172160.1 | - | 4 - 5 | 641 | 63 | |
AK220309.1 | - | 6 - 7 | 962 | 63 | |
BC024570.1 | - | 3 - 4 | 534 | 63 | |
BC039567.1 | - | 1 - 2 | 101 | 63 | |
BC043310.1 | - | 8 - 9 | 1373 | 63 |