Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plek
Entrez Gene ID
56193
Gene Name
pleckstrin
Gene Aliases
2010300B13Rik
Chromosome Location
Chr.11: 16971206 - 17008718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019549.2 NP_062422.1 3 - 4 500 95
GenBank mRNA AF073294.1 - 3 - 4 484 95
AF181829.1 - 3 - 4 444 95
AK033872.1 - 3 - 4 438 95
AK038804.1 - - 538 95
AK050340.1 - 3 - 4 442 95
AK075810.1 - 3 - 4 444 95
AK138831.1 - 3 - 4 438 95
AK151460.1 - 3 - 4 412 95
AK152501.1 - 3 - 4 412 95
AK153109.1 - 3 - 4 412 95
AK156235.1 - 3 - 4 501 95
BC066185.1 - 3 - 4 426 95

View Full Product Details