Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plek
Entrez Gene ID
56193
Gene Name
pleckstrin
Gene Aliases
2010300B13Rik
Chromosome Location
Chr.11: 16971206 - 17008718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019549.2 NP_062422.1 8 - 9 1040 79
GenBank mRNA AF073294.1 - 8 - 9 1024 79
AF181829.1 - 8 - 9 984 79
AF303745.1 - 9 - 10 972 79
AK033872.1 - 8 - 9 978 79
AK038804.1 - - 1078 79
AK050340.1 - 8 - 9 982 79
AK075810.1 - 8 - 9 984 79
AK138831.1 - 8 - 9 978 79
AK151460.1 - 8 - 9 952 79
AK152501.1 - 8 - 9 952 79
AK153109.1 - 8 - 9 952 79
AK156235.1 - 8 - 9 1041 79
BC066185.1 - 8 - 9 966 79

View Full Product Details