Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atad1
Entrez Gene ID
67979
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
4921525H23Rik, AW107648, Thorase
Chromosome Location
Chr.19: 32672558 - 32739766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026487.3 NP_080763.2 2 - 3 206 117
GenBank mRNA AK014967.1 - 2 - 3 223 117
AK030719.1 - 2 - 3 226 117
AK033639.1 - 2 - 3 230 117
AK044925.1 - 2 - 3 206 117
AK075685.1 - 2 - 3 250 117
AK150469.1 - 2 - 3 226 117
AK152059.1 - 2 - 3 226 117
AK165953.1 - 2 - 3 241 117
BC029085.1 - 2 - 3 206 117
BC043051.1 - 2 - 3 191 117

View Full Product Details