Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atad1
Entrez Gene ID
67979
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
4921525H23Rik, AW107648, Thorase
Chromosome Location
Chr.19: 32672558 - 32739766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026487.3 NP_080763.2 3 - 4 307 155
GenBank mRNA AK014967.1 - 3 - 4 324 155
AK030719.1 - 3 - 4 327 155
AK033639.1 - 3 - 4 331 155
AK044925.1 - 3 - 4 307 155
AK075685.1 - 3 - 4 351 155
AK150469.1 - 3 - 4 327 155
AK152059.1 - 3 - 4 327 155
AK165953.1 - 3 - 4 342 155
BC029085.1 - 3 - 4 307 155
BC043051.1 - 3 - 4 292 155

View Full Product Details