The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081150.1 | NP_001074619.1 | 11 - 12 | 2395 | 62 |
GenBank mRNA | BC094431.1 | - | 8 - 9 | 1202 | 62 |