Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc64
Entrez Gene ID
75665
Gene Name
coiled-coil domain containing 64
Gene Aliases
2210403N09Rik, BICDR-1
Chromosome Location
Chr.5: 115648174 - 115731643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK041990.1 - 2 - 3 446 80

View Full Product Details