Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rimbp2
Entrez Gene ID
231760
Gene Name
RIMS binding protein 2
Gene Aliases
A930033C01Rik, mKIAA0318
Chromosome Location
Chr.5: 128757791 - 128953647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081388.1 NP_001074857.1 20 - 21 3676 124
GenBank mRNA AK038814.1 - 19 - 20 3144 124
AK044685.1 - 19 - 20 3776 124
AK122245.2 - 11 - 12 2489 124
AK147417.1 - 16 - 17 3082 124
BC151100.1 - 16 - 17 3267 124
BC151101.1 - 16 - 17 3267 124

View Full Product Details