The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163471.1 | NP_001156943.1 | 3 - 4 | 544 | 63 |
NM_172637.3 | NP_766225.2 | 3 - 4 | 544 | 63 | |
GenBank mRNA | AK029549.1 | - | 3 - 4 | 404 | 63 |
AK041629.1 | - | 3 - 4 | 515 | 63 | |
AK043646.1 | - | 3 - 4 | 505 | 63 | |
AK079321.1 | - | 3 - 4 | 500 | 63 | |
BC137671.1 | - | 3 - 4 | 515 | 63 | |
BC144815.1 | - | 3 - 4 | 515 | 63 |